missing translation for 'onlineSavingsMsg'
Learn More

1-dodecylimidazole, 97%

Product Code. 10066952 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10066952

Brand: Thermo Scientific Acros 381700010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4303-67-7 | C15H28N2 | 236.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4303-67-7
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Boiling Point 146.0°C (0.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C15H28N2
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
MDL Number MFCD00278859
Quantity 1 g
Synonym 1-dodecyl-1h-imidazole, n-dodecylimidazole, 1h-imidazole, 1-dodecyl, n-laurylimidazole, imidazole, 1-dodecyl, 1-laurylimidazole, unii-q11nxv80s7, 1-dodecyl-1h-imidazol, n-dodecyl imidazole, pubchem20091
InChI Key JMTFLSQHQSFNTE-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCN1C=CN=C1
IUPAC Name 1-dodecylimidazole
Molecular Weight (g/mol) 236.40
PubChem CID 78002
Formula Weight 236.4
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Dodecylimidazole
Show More Show Less
compliance-icons
Product Identifier
  • 1-Dodecylimidazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.