missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne, 97%, Thermo Scientific Chemicals

Catalog Number 11337915
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
765-03-7
C12H22
166.31
MFCD00008960
ZVDBUOGYYYNMQI-UHFFFAOYSA-N
1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
69821
dodec-1-yne
CCCCCCCCCCC#C
This item is not returnable. View return policy
765-03-7
C12H22
166.31
MFCD00008960
ZVDBUOGYYYNMQI-UHFFFAOYSA-N
1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
69821
dodec-1-yne
CCCCCCCCCCC#C

1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Solubility
Not miscible in water.

Notes
Incompatible with heavy metal powders and oxidizing agents. Store between 2 - 8°C. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -19°C
Density 0.779
Boiling Point 214°C to 216°C
Flash Point 79°C (174°F)
Refractive Index 1.434
Quantity 5 g
Beilstein 1741320
Solubility Information Not miscible in water.
Formula Weight 166.31
Percent Purity 97%
Chemical Name or Material 1-Dodecyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'