missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanol, 96%

Product Code. 11332726
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11332726

Brand: Thermo Scientific Alfa Aesar L08807.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-96-9 | C20H42O | 298.56 g/mol

1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol?s high molecular weight alcohols (policosanol?s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol′s high molecular weight alcohols (policosanol′s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-96-9
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.56
MDL Number MFCD00002938
InChI Key BTFJIXJJCSYFAL-UHFFFAOYSA-N
Synonym 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
PubChem CID 12404
ChEBI CHEBI:75627
IUPAC Name icosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 62°C to 66°C
Density 0.841
Boiling Point 220°C to 225°C (3 mmHg)
Flash Point 195°C (383°F)
Quantity 100 g
Beilstein 1705104
Solubility Information Insoluble in water.
Percent Purity 96%
Chemical Name or Material 1-Eicosanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.