missing translation for 'onlineSavingsMsg'
Learn More

1-Eth-1-ynyl-4-hexylbenzene, 97%, Thermo Scientific™

Product Code. 10173444
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 79887-11-9
Molecular Formula C14H18
Molecular Weight (g/mol) 186.30
MDL Number MFCD00173883
InChI Key NFPDFDTYANKKIU-UHFFFAOYSA-N
Synonym 1-eth-1-ynyl-4-hexylbenzene, 4-hexyl-1-ethynylbenzene, 4-hexylphenylacetylene, 1-ethynyl-4-hexyl-benzene, 4-ethynyl-1-hexylbenzene, acmc-209pie, 4-n-hexylphenylacetylene, 1-ethynyl-4-n-hexylbenzene
PubChem CID 2775129
IUPAC Name 1-ethynyl-4-hexylbenzene
SMILES CCCCCCC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HCidentical toC6H4(CH2)5CH3
Quantity 25 g
Formula Weight 186.3
Percent Purity 97%
Chemical Name or Material 1-Eth-1-ynyl-4-hexylbenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'