missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylethyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Product Code. 11449943
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30018-16-7
Molecular Formula C23H24BrO2P
Molecular Weight (g/mol) 443.32
MDL Number MFCD00031605
InChI Key RSYXORMKBUFAMS-UHFFFAOYNA-M
Synonym 1-ethoxy-1-oxopropan-2-yl triphenylphosphonium bromide, 1-ethoxycarbonyl ethyltriphenylphosphonium bromide, 1-ethoxycarbonyl ethyl triphenylphosphonium bromide, 2-ethoxy-1-methyl-2-oxoethyl triphenylphosphonium bromide, ceetppb, 1-carbethoxyethyl triphenylphosphonium bromide, 1-ethoxycarbonylethyl triphenylphosphonium bromide, 1-ethoxy-1-oxopropan-2-yl triphenylphosphanium bromide, phosphonium, 2-ethoxy-1-methyl-2-oxoethyl triphenyl-, bromide, ethoxycarbonylethyltriphenylphosphoniumbromide
PubChem CID 3084511
IUPAC Name (1-ethoxy-1-oxopropan-2-yl)-triphenylphosphanium;bromide
SMILES [Br-].CCOC(=O)C(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼130°C (decomposition)
Quantity 500 g
Beilstein 3822244
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 443.33
Percent Purity 97%
Chemical Name or Material (1-Ethoxycarbonylethyl)triphenylphosphonium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'