missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-1-methylpyrrolidinium bromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Specifications
Quantity | 25 g |
Beilstein | 14507266 |
Solubility Information | Soluble in water. |
Formula Weight | 194.12 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 1-Ethyl-1-methylpyrrolidinium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'