Learn More
1-Ethyl-2-methylquinolinium iodide, 97%, Thermo Scientific Chemicals
£37.80 - £317.00
Chemical Identifiers
CAS | 606-55-3 |
---|---|
Molecular Formula | C12H14IN |
Molecular Weight (g/mol) | 299.155 |
MDL Number | MFCD00031829 |
InChI Key | OEVSHJVOKFWBJY-UHFFFAOYSA-M |
Synonym | 1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1 |
PubChem CID | 69076 |
IUPAC Name | 1-ethyl-2-methylquinolin-1-ium;iodide |
SMILES | CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473244
|
Thermo Scientific Alfa Aesar
A10584.06 |
5 g |
£37.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483244
|
Thermo Scientific Alfa Aesar
A10584.14 |
25 g |
£113.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493244
|
Thermo Scientific Alfa Aesar
A10584.22 |
100 g |
£317.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
Solubility
Soluble in Water.
Notes
Precursor in synthesis of carbocyanine dyes.
Chemical Identifiers
606-55-3 | |
299.155 | |
OEVSHJVOKFWBJY-UHFFFAOYSA-M | |
69076 | |
CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-] |
C12H14IN | |
MFCD00031829 | |
1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1 | |
1-ethyl-2-methylquinolin-1-ium;iodide |
Specifications
606-55-3 | |
C12H14IN | |
Light sensitive | |
Soluble in Water. | |
CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-] | |
299.155 | |
299.16 | |
1-Ethyl-2-methylquinolinium iodide |
231°C (decomposition) | |
MFCD00031829 | |
1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1 | |
OEVSHJVOKFWBJY-UHFFFAOYSA-M | |
1-ethyl-2-methylquinolin-1-ium;iodide | |
69076 | |
97% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 210-120-7
missing translation for 'rtecsNumber' : VA0740000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only