missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2-pyrrolidinone, 98%

Product Code. 10658244 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10658244

Brand: Thermo Scientific Acros 163935000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2687-91-4 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-91-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00003199
InChI Key ZFPGARUNNKGOBB-UHFFFAOYSA-N
Synonym 1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone, n-ethyl-2-pyrrolidinone
PubChem CID 17595
IUPAC Name 1-ethylpyrrolidin-2-one
SMILES CCN1CCCC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -75.0°C
Density 0.9980g/mL
Boiling Point 212.5°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 500 g
Fieser 09,220
Specific Gravity 0.998
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Viscosity 2.1 mPa.s (20°C)
Formula Weight 113.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Ethyl-2-pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Ethyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.