Learn More
1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals
£82.10 - £445.00
Chemical Identifiers
CAS | 143314-17-4 |
---|---|
Molecular Formula | C8H14N2O2 |
Molecular Weight (g/mol) | 170.212 |
MDL Number | MFCD06798186 |
InChI Key | XIYUIMLQTKODPS-UHFFFAOYSA-M |
Synonym | 1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc |
PubChem CID | 11658353 |
IUPAC Name | 1-ethyl-3-methylimidazol-3-ium;acetate |
SMILES | CCN1C=C[N+](=C1)C.CC(=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15475538
|
Thermo Scientific Alfa Aesar
H59277.06 |
5 g |
£82.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15485538
|
Thermo Scientific Alfa Aesar
H59277.18 |
50 g |
£445.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a solvent and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a solvent and as pharmaceutical intermediate.
Solubility
Soluble in water, acetonitrile, acetone, dichloromethane.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
143314-17-4 | |
170.212 | |
XIYUIMLQTKODPS-UHFFFAOYSA-M | |
11658353 | |
CCN1C=C[N+](=C1)C.CC(=O)[O-] |
C8H14N2O2 | |
MFCD06798186 | |
1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc | |
1-ethyl-3-methylimidazol-3-ium;acetate |
Specifications
143314-17-4 | |
1.027 g/mL | |
C8H14N2O2 | |
MFCD06798186 | |
1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc | |
XIYUIMLQTKODPS-UHFFFAOYSA-M | |
1-ethyl-3-methylimidazol-3-ium;acetate | |
11658353 | |
97% | |
1-Ethyl-3-methylimidazolium acetate |
-20°C | |
164°C (327°F) | |
n20/D 1.502 | |
Moisture sensitive | |
Soluble in water,acetonitrile,acetone,dichloromethane. | |
CCN1C=C[N+](=C1)C.CC(=O)[O-] | |
170.212 | |
170.21 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only