missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethylcyclohexanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 0.9190g/mL |
Boiling Point | 166.0°C |
Flash Point | 68°C |
Assay Percent Range | 96% min. (GC) |
Quantity | 5 mL |
Specific Gravity | 0.919 |
Formula Weight | 128.22 |
Percent Purity | 97% |
Chemical Name or Material | 1-Ethylcyclohexanol |
Safety and Handling
Product Identifier
- 1-Ethylcyclohexanol, 97%
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'