missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylcyclohexanol, 97%, Thermo Scientific Chemicals

Product Code. 15289856
Click to view available options
Quantity:
5 mL
25 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1940-18-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00021402
InChI Key BUCJHJXFXUZJHL-UHFFFAOYSA-N
Synonym 1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
PubChem CID 16021
IUPAC Name 1-ethylcyclohexan-1-ol
SMILES CCC1(O)CCCCC1

Specifications

Density 0.9190g/mL
Boiling Point 166.0°C
Flash Point 68°C
Assay Percent Range 96% min. (GC)
Quantity 25 mL
Specific Gravity 0.919
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material 1-Ethylcyclohexanol
compliance-icons
Product Identifier
  • 1-Ethylcyclohexanol, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'