missing translation for 'onlineSavingsMsg'
Learn More
1-Ethylpiperidine, 99%, Thermo Scientific Chemicals Product Code.: 10132530

1-Ethylpiperidine, 99%, Thermo Scientific Chemicals

Product Code. 10132530
500 mL, Glass bottle
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10132530

Brand: Thermo Scientific Acros 118885000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-09-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.2
MDL Number MFCD00006507
InChI Key HTLZVHNRZJPSMI-UHFFFAOYSA-N
Synonym n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene
PubChem CID 13007
ChEBI CHEBI:39017
IUPAC Name 1-ethylpiperidine
SMILES CCN1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20.0°C
Color Colorless to Yellow
Density 0.8240g/mL
Boiling Point 129.0°C to 131.0°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.4450
Quantity 500 mL
Beilstein 20,17
Specific Gravity 0.824
Solubility Information Solubility in water: 50g/L (20°C)
Formula Weight 113.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Ethylpiperidine, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Ethylpiperidine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.