missing translation for 'onlineSavingsMsg'
Learn More
1-Ethynyl-1-cyclohexanol, 99+%, Thermo Scientific Chemicals Product Code.: 10569600

1-Ethynyl-1-cyclohexanol, 99+%, Thermo Scientific Chemicals

Product Code. 10569600
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

Product Code. 10569600

Brand: Thermo Scientific Acros 119101000

Please to purchase this item. Need a web account? Register with us today!

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-27-3
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00003858
InChI Key QYLFHLNFIHBCPR-UHFFFAOYSA-N
Synonym 1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol
PubChem CID 6525
IUPAC Name 1-ethynylcyclohexan-1-ol
SMILES C#CC1(CCCCC1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 30.0°C to 32.0°C
Color Colorless to Yellow
Density 0.9763g/mL
Boiling Point 180.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CC6H10OH
Refractive Index 1.4810 to 1.4840
Quantity 100 g
Beilstein 06,II,1
Merck Index 10,381
Specific Gravity 0.9763
Solubility Information Solubility in water: 24.5g/L (20°C). Other solubilities: soluble in most organic solvents, soluble in diethyl ether, alcohols, esters, ketone, and chlorinated hydrocarbons
Formula Weight 124.18
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material 1-Ethynyl-1-cyclohexanol, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453256 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number119105000,119100025,119101000
Lot NumberA0402934 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
Lot NumberA0403273 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
Lot NumberA0398433 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
Lot NumberA0396538 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
compliance-icons
Product Identifier
  • 1-Ethynyl-1-cyclohexanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Ethynyl-1-cyclohexanol, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.