missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-2-fluorobenzene, 97%, Thermo Scientific Chemicals

Product Code. 10576363
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-49-4
Molecular Formula C8H5F
Molecular Weight (g/mol) 120.13
MDL Number MFCD00799540
InChI Key YFPQIXUNBPQKQR-UHFFFAOYSA-N
Synonym 2-fluorophenylacetylene, 2'-fluorophenyl acetylene, 1-ethynyl-2-fluoro-benzene, 2'-fluorophenylacetylene, 2-fluorophenylethyne, o-fluorophenylacetylene, benzene, ethynylfluoro, 2-fluoro-phenylacetylene, 2-fluorophenyl-ethyne, 2-fluorophenyl acetylene
PubChem CID 643358
IUPAC Name 1-ethynyl-2-fluorobenzene
SMILES FC1=CC=CC=C1C#C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.0600g/mL
Boiling Point 150°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4C∽CH
Refractive Index 1.5240 to 1.527
Quantity 1 g
Specific Gravity 1.06
Formula Weight 120.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Ethynyl-2-fluorobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Ethynyl-2-fluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'