Learn More
1-Ethynylcyclohexanol, 99%, Thermo Scientific Chemicals
£29.20 - £103.00
Chemical Identifiers
CAS | 78-27-3 |
---|---|
Molecular Formula | C8H12O |
Molecular Weight (g/mol) | 124.183 |
MDL Number | MFCD00003858 |
InChI Key | QYLFHLNFIHBCPR-UHFFFAOYSA-N |
Synonym | 1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol |
PubChem CID | 6525 |
IUPAC Name | 1-ethynylcyclohexan-1-ol |
SMILES | C#CC1(CCCCC1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323256
|
Thermo Scientific Alfa Aesar
L12591.22 |
100 g |
£29.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333256
|
Thermo Scientific Alfa Aesar
L12591.36 |
500 g |
£103.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Ethynylcyclohexanol is used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Ethynylcyclohexanol is used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water (10g/L at 20°C), diethyl ether, alcohols, esters, and ketone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
78-27-3 | |
124.183 | |
QYLFHLNFIHBCPR-UHFFFAOYSA-N | |
6525 | |
C#CC1(CCCCC1)O |
C8H12O | |
MFCD00003858 | |
1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol | |
1-ethynylcyclohexan-1-ol |
Specifications
78-27-3 | |
0.973 | |
73°C (163°F) | |
C8H12O | |
MFCD00003858 | |
471404 | |
Soluble in water (10g/L at 20°C),diethyl ether,alcohols,esters,and ketone. | |
C#CC1(CCCCC1)O | |
124.183 | |
124.18 | |
1-Ethynylcyclohexanol |
30°C to 33°C | |
180°C | |
Mild | |
1.483 | |
UN2811 | |
1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol | |
QYLFHLNFIHBCPR-UHFFFAOYSA-N | |
1-ethynylcyclohexan-1-ol | |
6525 | |
99% |
Safety and Handling
GHS H Statement
H311-H302-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H302-H311-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 201-100-9
missing translation for 'rtecsNumber' : GV9100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only