missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylcyclopentanol, 98+%, Thermo Scientific Chemicals

Product Code. 11496316
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Ethynylcyclopentanol is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylcyclopentanol is used in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17356-19-3
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00001365
InChI Key LQMDOONLLAJAPZ-UHFFFAOYSA-N
Synonym 1-ethynylcyclopentanol, cyclopentanol, 1-ethynyl, 1-ethynyl-1-cyclopentanol, cyclopentyl ethynyl carbinol, 1-ethynyl-cyclopentan-1-ol, acmc-20abvz, 1-ethinylcyclopentanol, 1-ethynyl-cyclopentanol, cyclopentanol, ethynyl, 1-ethynylcyclopentanol ?
PubChem CID 87074
IUPAC Name 1-ethynylcyclopentan-1-ol
SMILES C#CC1(CCCC1)O

Specifications

Melting Point 23°C to 27°C
Density 0.962
Boiling Point 156°C to 159°C
Flash Point 48°C (118°F)
Odor Irritating
Refractive Index 1.474
Quantity 100 g
UN Number UN1325
Beilstein 1924167
Solubility Information Insoluble in water.
Formula Weight 110.16
Percent Purity ≥98%
Chemical Name or Material 1-Ethynylcyclopentanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'