missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Fluoro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.
Solubility
Soluble in chloroform, benzene, ether and propylene glycol.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.
Solubility
Soluble in chloroform, benzene, ether and propylene glycol.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | 24°C to 30°C |
Density | 1.482 |
Boiling Point | 296°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.569 |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 398632 |
Merck Index | 14,4172 |
Solubility Information | Soluble in chloroform,benzene,ether and propylene glycol. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'