missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-(trifluoromethylsulfonyl)benzene, 95%, Thermo Scientific™

Product Code. 15494946
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2358-41-0
Molecular Formula C7H4F4O2S
Molecular Weight (g/mol) 228.161
MDL Number MFCD09971596
InChI Key NXZFXDLCZKKNCX-UHFFFAOYSA-N
Synonym 1-fluoro-2-trifluoromethyl sulfonyl benzene, 1-fluoro-2-trifluoromethylsulfonyl benzene, 1-fluoro-2-trifluoromethanesulfonylbenzene, o-fluorophenyl trifluoromethyl sulfone, 2-fluorophenyl trifluoromethyl sulfone, 1-fluoro-2-trifluoromethylsulfonyl-benzene, 1-fluoro-2-trifluoromethane sulfonylbenzene, 1-fluoro-2-trifluoromethanesulfonyl benzene, 1-fluoro-2-trifluoromethyl ;sulfonyl ;benzene
PubChem CID 15731661
IUPAC Name 1-fluoro-2-(trifluoromethylsulfonyl)benzene
SMILES C1=CC=C(C(=C1)F)S(=O)(=O)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 228.16
Percent Purity 95%
Chemical Name or Material 1-Fluoro-2-(trifluoromethylsulfonyl)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'