missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-iodobenzene, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10246690
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-86-4
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001044
InChI Key VSKSBSORLCDRHS-UHFFFAOYSA-N
Synonym 3-fluoroiodobenzene, m-fluoroiodobenzene, 3-iodofluorobenzene, m-iodofluorobenzene, benzene, 1-fluoro-3-iodo, m-fluorobenzene, 1-fluoro-3-iodo-benzene, 1-iodo-3-fluorobenzene, 3-fluoro-1-iodobenzene, m-fluorjodbenzol
PubChem CID 70725
IUPAC Name 1-fluoro-3-iodobenzene
SMILES FC1=CC=CC(I)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow or Pink
Density 1.8900g/mL
Boiling Point 77°C to 78°C (19.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4I
Refractive Index 1.5830 to 1.585
Quantity 5 g
Beilstein 05, III, 578
Specific Gravity 1.89
Formula Weight 222
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-3-iodobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Fluoro-3-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'