Learn More
1-(Fmoc-amino)cyclohexanecarboxylic acid, 98+%, Thermo Scientific Chemicals
Description
1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 185°C to 187°C (decomposition) |
Quantity | 1 g |
Beilstein | 7887646 |
Solubility Information | Insoluble in water. |
Formula Weight | 365.43 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-(Fmoc-amino)cyclohexanecarboxylic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q13H005 | Certificate of Analysis | 06/12/2021 | L19810.06,L19810.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.