missing translation for 'onlineSavingsMsg'
Learn More

1-Fmoc-piperidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15430879
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148928-15-8
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD00273475
InChI Key OJYOOHSQOIDDOO-UHFFFAOYSA-N
Synonym fmoc-inp-oh, fmoc-isonipecotic acid, 1-fmoc-piperidine-4-carboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl piperidine-4-carboxylic acid, n-fmoc-isonipecotic acid, fmoc-piperidine-4-carboxylic acid, n-fmoc-4-piperidinecarboxylic acid, 1,4-piperidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl ester, n-9-fluorenylmethyloxycarbonyl-isonipecotic acid, 1-9h-fluoren-9-ylmethoxycarbonyl piperidine-4-carboxylic acid
PubChem CID 2736490
IUPAC Name 1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point ∼190°C (decomposition)
Color White
Quantity 1 g
Beilstein 7722550
Formula Weight 351.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Fmoc-piperidine-4-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'