missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
Click to view available options
Quantity:
25 g
100 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical, Quantity: 100g, Packaging: Plastic powder jar, CAS: 22767-50-6, White, Melting Point: 300 deg.C, Molecular Formula: C7H15NaO3S, Molecular Weight: 202.244
Specifications
Specifications
Melting Point | 300°C |
Color | White |
Physical Form | Powder/Solid |
Quantity | 5 kg |
Formula Weight | 202.25g/mol |
Packaging | Plastic keg |
Safety and Handling
Product Identifier
- 1-Heptanesulfonic acid, sodium salt
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'