missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade

Product Code. 10107330 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10107330

Brand: Thermo Scientific undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207300-90-1 | C7H15NaO3S·H2O | 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207300-90-1
InChI Key XWZCREJRXRKIRQ-UHFFFAOYSA-M
Synonym sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
PubChem CID 23687711
IUPAC Name sodium;heptane-1-sulfonate;hydrate
SMILES CCCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Absorbance (2.7g in 100mL water) (pH 3), (2.7 g in 100 ml water) (pH 3) at 0nm, 0.06 max. at 254nm, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm
Melting Point >300°C
Color White
Loss on Drying 7.4% to 9% (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Packaging Glass Bottle
Molecular Formula C7H17NaO4S
Linear Formula CH3(CH2)6SO3Na·H2O
Quantity 25 g
Solubility Information Solubility in water: Soluble
Molecular Weight (g/mol) 220.259
Formula Weight 220.27
Physical Form Powder or Flakes
Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt Monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • 1-Heptanesulfonic acid, sodium salt monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Heptanesulfonic acid sodium salt monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.