missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanol, 98%, Thermo Scientific Chemicals

£23.72 - £84.92

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419123
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11180.36
500 g
£23.72
500g
Please to purchase this item. Need a web account? Register with us today!
11429123
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11180.0E
2500 g
£84.92
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in alcohol, chloroform, ether

Notes
Keep away from direct sunlight. Incompatible with strong oxidizing agents, strong acids. Store at 2-8°C.
Specifications

Chemical Identifiers

36653-82-4
242.45
BXWNKGSJHAJOGX-UHFFFAOYSA-N
2682
hexadecan-1-ol
C16H34O
MFCD00004760
1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
CHEBI:16125
CCCCCCCCCCCCCCCCO

Specifications

36653-82-4
0.817 g/mL
135°C (275°F)
C16H34O
1748475
1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
BXWNKGSJHAJOGX-UHFFFAOYSA-N
hexadecan-1-ol
2682
242.45
1-Hexadecanol
47°C to 51°C
190°C (15 mm)
98%
MFCD00004760
14,2028
Soluble in alcohol, chloroform, ether
CCCCCCCCCCCCCCCCO
242.45
CHEBI:16125
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 253-149-0

missing translation for 'rtecsNumber' : MM0225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-Hexadecanol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only