missing translation for 'onlineSavingsMsg'
Learn More
(1-Hexadecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals Product Code.: 11478740

(1-Hexadecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals

Product Code. 11478740
100 g
Click to view available options
Quantity:
100 g
500 g
5000 g
This item is not returnable. View return policy

Product Code. 11478740

Brand: Thermo Scientific Alfa Aesar A15235.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(1-Hexadecyl)trimethylammonium bromide is a cationic surfactant in hair conditioning products. It acts as a phase-transfer catalyst for the preparation of polyamide, polycarbonate and polythiocarbonate. It facilitates ligand-free palladium salt-catalyzed Heck and Suzuki cross-coupling reactions in water. It is an active component of the topical antiseptic cetrimide and used in household products such as shampoos and cosmetics. As a buffer solution, it is involved in the extraction of DNA as well as in the synthesis of gold nanoparticles and silican nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexadecyl)trimethylammonium bromide is a cationic surfactant in hair conditioning products. It acts as a phase-transfer catalyst for the preparation of polyamide, polycarbonate and polythiocarbonate. It facilitates ligand-free palladium salt-catalyzed Heck and Suzuki cross-coupling reactions in water. It is an active component of the topical antiseptic cetrimide and used in household products such as shampoos and cosmetics. As a buffer solution, it is involved in the extraction of DNA as well as in the synthesis of gold nanoparticles and silican nanoparticles.

Solubility
Soluble in alcohol, chloroform and water. Slightly soluble in acetone. Insoluble in diethyl ether and benzene.

Notes
Hygroscopic. Store in an airtight container. Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
PubChem CID 5974
ChEBI CHEBI:3567
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point ∼245°C (decomposition)
pH 5 to 7
Flash Point 244°C (471°F)
Odor Odorless
Quantity 100 g
UN Number UN3077
Beilstein 3598189
Sensitivity Hygroscopic
Merck Index 14,2025
Solubility Information Soluble in alcohol,chloroform and water. Slightly soluble in acetone. Insoluble in diethyl ether and benzene.
IUPAC Name hexadecyl(trimethyl)azanium;bromide
Formula Weight 364.46
Percent Purity 98%
Chemical Name or Material (1-Hexadecyl)trimethylammonium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (1-Hexadecyl)trimethylammonium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1-Hexadecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.