missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals

Product Code. 11314837
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(1-Hexadecyl)trimethylammonium chloride is used as a topical antiseptic and surfactant. It is used in hair care formulations, cream rinse ingredient, hair conditioners and shampoos as a conditioning agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-02-7
Molecular Formula C19H42ClN
Molecular Weight (g/mol) 320.00
MDL Number MFCD00011773
InChI Key WOWHHFRSBJGXCM-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium chloride, cetrimonium chloride, cetyltrimethylammonium chloride, n,n,n-trimethylhexadecan-1-aminium chloride, dehyquart a, cetyl trimethyl ammonium chloride, aliquat 6, trimethylcetylammonium chloride, arquad 16-50, n-hexadecyltrimethylammonium chloride
PubChem CID 8154
ChEBI CHEBI:53581
IUPAC Name hexadecyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 234°C
Color White
Density 0.968 g/mL
Odor Odorless
Linear Formula CH3(CH2)15N(CH3)3Cl
Quantity 5 g
UN Number UN1759
Beilstein 3657974
Solubility Information Soluble in water.
Formula Weight 320
Percent Purity 96%
Physical Form Powder
Chemical Name or Material (1-Hexadecyl)trimethylammonium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'