Learn More
1-Hexadecylamine, 90%, Thermo Scientific Chemicals
£15.50 - £133.00
Chemical Identifiers
CAS | 143-27-1 |
---|---|
Molecular Formula | C16H35N |
Molecular Weight (g/mol) | 241.46 |
MDL Number | MFCD00008158 |
InChI Key | FJLUATLTXUNBOT-UHFFFAOYSA-N |
Synonym | hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine |
PubChem CID | 8926 |
IUPAC Name | hexadecan-1-amine |
SMILES | CCCCCCCCCCCCCCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011070
|
Thermo Scientific Acros
120510050 |
5 g | Glass bottle |
£16.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10724241
|
Thermo Scientific Acros
120511000 |
100 g | Glass bottle |
£15.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10112630
|
Thermo Scientific Acros
120515000 |
500 g | Glass bottle |
£37.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11955171
|
Thermo Scientific Acros
120510025 |
2.5 kg | Plastic drum |
£133.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
143-27-1 | |
241.46 | |
FJLUATLTXUNBOT-UHFFFAOYSA-N | |
8926 | |
CCCCCCCCCCCCCCCCN |
C16H35N | |
MFCD00008158 | |
hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine | |
hexadecan-1-amine |
Specifications
143-27-1 | |
White | |
140°C | |
88% min. (GC) | |
CH3(CH2)15NH2 | |
04,202 | |
Solubility in water: insoluble | |
CCCCCCCCCCCCCCCCN | |
241.46 | |
241.46 | |
Low Melting Crystalline Mass or Chunks |
38.0°C to 47.0°C | |
330.0°C | |
Authentic | |
C16H35N | |
MFCD00008158 | |
hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine | |
FJLUATLTXUNBOT-UHFFFAOYSA-N | |
hexadecan-1-amine | |
8926 | |
90% | |
1-Hexadecylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-596-8
- 1-Hexadecylamine, straight-chained C16
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P391-Collect spillage.
RUO – Research Use Only