missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecylamine, tech. 90%, remainder mainly 1-octadecylamine, Thermo Scientific Chemicals

Product Code. 11448740
Click to view available options
Quantity:
25 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-27-1
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00008158
InChI Key FJLUATLTXUNBOT-UHFFFAOYSA-N
Synonym hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine
PubChem CID 8926
IUPAC Name hexadecan-1-amine
SMILES CCCCCCCCCCCCCCCCN

Specifications

Melting Point 42°C to 47°C
Density 0.813
Boiling Point 144°C (2 mmHg)
Flash Point 140°C (284°F)
Assay Percent Range 90%
Refractive Index 1.4496
Quantity 25 g
UN Number UN3259
Beilstein 1634065
Solubility Information Insoluble in water.
Formula Weight 241.46
Percent Purity 90%
Grade Technical
Assay remainder mainly 1-octadecylamine
Chemical Name or Material 1-Hexadecylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'