missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecyne, tech. 90%, Thermo Scientific Chemicals

Product Code. 11470173
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-74-3
Molecular Formula C16H30
Molecular Weight (g/mol) 222.42
MDL Number MFCD00015084
InChI Key UCIDYSLOTJMRAM-UHFFFAOYSA-N
Synonym 1-hexadecyne, tetradecylacetylene, acmc-209nbc
PubChem CID 12396
IUPAC Name hexadec-1-yne
SMILES CCCCCCCCCCCCCCC#C

Specifications

Melting Point 13°C to 16°C
Density 0.79
Boiling Point 146°C to 148°C (10 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)13C≡CH
Refractive Index 1.442
Quantity 5 g
Beilstein 1702804
Formula Weight 222.42
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Hexadecyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'