missing translation for 'onlineSavingsMsg'
Learn More

1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, Certified, Fisher Chemical™

1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, Certified, Fisher Chemical, Quantity: 100g, CAS: 2832-45-3, White, Melting Point: 300 deg.C, Molecular Formula: C6H13NaO3S, Molecular Weight: 188.217, Packaging: Plastic powder jar

£189.00 - £556.00

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
SMILES [Na+].CCCCCCS([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10666642
View Documents
Fisher Chemical
H/0438/46
25 g
£189.00
25g
Please to purchase this item. Need a web account? Register with us today!
10356230
View Documents Promotion Details
Fisher Chemical
H/0438/48
100 g
£556.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, Certified, Fisher Chemical, Quantity: 100g, CAS: 2832-45-3, White, Melting Point: 300 deg.C, Molecular Formula: C6H13NaO3S, Molecular Weight: 188.217, Packaging: Plastic powder jar

Specifications

Chemical Identifiers

2832-45-3
188.22
QWSZRRAAFHGKCH-UHFFFAOYSA-M
23677630
C6H13NaO3S
MFCD00007542
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
[Na+].CCCCCCS([O-])(=O)=O

Specifications

300°C
White
C6H13NaO3S
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
[Na+].CCCCCCS([O-])(=O)=O
188.22
188.22g/mol
2832-45-3
Powder/Solid
MFCD00007542
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium hexane-1-sulfonate
23677630
Plastic powder jar
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Hexanesulfonic acid, sodium salt

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.