Learn More
1-hexanesulfonic acid, sodium salt hydrate, 99%, Thermo Scientific Chemicals
£36.00 - £364.00
Chemical Identifiers
CAS | 2832-45-3 |
---|---|
Molecular Formula | C6H13NaO3S |
Molecular Weight (g/mol) | 188.22 |
MDL Number | MFCD00007542 |
InChI Key | QWSZRRAAFHGKCH-UHFFFAOYSA-M |
Synonym | sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt |
PubChem CID | 23677630 |
IUPAC Name | sodium;hexane-1-sulfonate |
SMILES | [Na+].CCCCCCS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10062740
|
Thermo Scientific Acros
206120050 |
5 g | Glass bottle |
£36.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10553951
|
Thermo Scientific Acros
206120250 |
25 g | Glass bottle |
£115.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10329233
|
Thermo Scientific Acros
206121000 |
100 g | Glass bottle |
£364.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2832-45-3 | |
188.22 | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
23677630 | |
[Na+].CCCCCCS([O-])(=O)=O |
C6H13NaO3S | |
MFCD00007542 | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
sodium;hexane-1-sulfonate |
Specifications
2832-45-3 | |
>300.0°C | |
Negligible | |
Authentic | |
C6H13NaO3S | |
MFCD00007542 | |
(10% in water) Clear colorless | |
[Na+].CCCCCCS([O-])(=O)=O | |
188.22 | |
188.22 | |
99% | |
1-Hexanesulfonic acid, sodium salt hydrate |
5.5 w/v% aq. soln., pH 3, H3PO4, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm, 5.5 w/v% aq. soln., pH 3, H3PO4 at 0nm | |
White | |
9.6% max. (100°C, 3 hrs) (vacuum) | |
99% | |
CH3(CH2)5SO3Na·xH2O | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
sodium;hexane-1-sulfonate | |
23677630 | |
Negligible | |
Crystalline Flakes or Solid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
RUO – Research Use Only