missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | -140.0°C |
Color | Colorless |
Density | 0.6780g/mL |
Boiling Point | 62.0°C to 65.0°C |
Flash Point | −26°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 1-Hexene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : 1-Hexene
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction