Learn More
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals
£62.50 - £632.00
Chemical Identifiers
CAS | 304680-35-1 |
---|---|
Molecular Formula | C10H19F6N2P |
Molecular Weight (g/mol) | 312.24 |
MDL Number | MFCD03093296 |
InChI Key | YPWSSSRXUOQNMQ-UHFFFAOYSA-N |
Synonym | 1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate |
PubChem CID | 2734175 |
IUPAC Name | 1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate |
SMILES | F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10371144
|
Thermo Scientific Acros
354180050 |
5 g | Plastic bottle |
£62.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041741
|
Thermo Scientific Acros
354180250 |
25 g | Plastic bottle |
£206.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10616744
|
Thermo Scientific Acros
354181000 |
100 g | Plastic bottle |
£632.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquidChemische Identifikatoren
304680-35-1 | |
312.24 | |
YPWSSSRXUOQNMQ-UHFFFAOYSA-N | |
2734175 | |
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1 |
C10H19F6N2P | |
MFCD03093296 | |
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate | |
1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate |
Spezifikation
304680-35-1 | |
100.0 | |
White | |
Authentic | |
C10H19F6N2P | |
MFCD03093296 | |
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate | |
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1 | |
312.24 | |
497 mPa.s (25°C) | |
99% | |
1-Hexyl-3-methylimidazolium hexafluorophosphate |
98.5 | |
-73.5°C | |
1.3045g/mL | |
99% | |
1.4160 to 1.418 | |
1.3045 | |
YPWSSSRXUOQNMQ-UHFFFAOYSA-N | |
1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate | |
2734175 | |
312.24 | |
Powder |
Sicherheit und Handhabung
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'tsca' : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
- 1-Hexyl-3-methylimidazolium hexafluorophosphate
Signalwort
- Warning
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only