missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxymethyl-5,5-dimethylhydantoin, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1-Hydroxymethyl-5,5-dimethylhydantoin is used as chelator for light-sensitive pharmaceutical drug, and as raw material of water-soluble polymers. It also can be used as raw material of disinfectants and preservatives, and soluble inhibitor for proteins. It is used as an adjuvant in the bleaching of food-contact recycled paper and board used in food packaging.
Solubility
Freely soluble in water.
Notes
Moisture Sensitive. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
1-Hydroxymethyl-5,5-dimethylhydantoin is used as chelator for light-sensitive pharmaceutical drug, and as raw material of water-soluble polymers. It also can be used as raw material of disinfectants and preservatives, and soluble inhibitor for proteins. It is used as an adjuvant in the bleaching of food-contact recycled paper and board used in food packaging.
Solubility
Freely soluble in water.
Notes
Moisture Sensitive. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Specifications
Melting Point | 104°C to 106°C |
Quantity | 100 g |
Beilstein | 139160 |
Merck Index | 14,4831 |
Solubility Information | Freely soluble in water. |
Formula Weight | 158.16 |
Percent Purity | 97% |
Chemical Name or Material | 1-Hydroxymethyl-5,5-dimethylhydantoin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'