missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-^m-hydrotetracarbonyldiruthenium(II), 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
Description
1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-μ-hydroteracarbonyldiruthenium(II) is widely used in various types of applications in organic synthesis. It is used for hydrogenation of carbonyls, alkenes and imines. It is also used in the alkylation of amines and amination of propargyl alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Calayst for hydrogen transfers including reduction of ketones and aldehydes, and oxidation and/or racemization of amines and alcohols
Specifications
Specifications
Melting Point | 223°C to 227°C |
Color | Yellow to Orange |
Quantity | 0.5 g |
Sensitivity | Moisture sensitive |
Merck Index | 14,8483 |
Solubility Information | Soluble in water and polar organic solvents. |
Formula Weight | 1085.15 |
Percent Purity | 98% |
Physical Form | Crystalline |
Chemical Name or Material | 1-Hydroxytetraphenylcyclopentadienyl (tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldir |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'