missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Indanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
50 g
Description
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.
Solubility
Soluble in water 10 g/L at 20°C, ethanol, benzene.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.
Solubility
Soluble in water 10 g/L at 20°C, ethanol, benzene.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 50°C to 54°C |
Boiling Point | 128°C (12 mmHg) |
Flash Point | 145°C (293°F) |
Quantity | 5 g |
Beilstein | 2042960 |
Solubility Information | Soluble in water 10g/L at 20°C,ethanol,benzene. |
Formula Weight | 134.18 |
Percent Purity | 98% |
Chemical Name or Material | 1-Indanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'