missing translation for 'onlineSavingsMsg'
Learn More

1-Indanone, 99+%, Thermo Scientific Chemicals

£27.20 - £535.93

Chemical Identifiers

CAS 83-33-0
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
InChI Key QNXSIUBBGPHDDE-UHFFFAOYSA-N
Synonym 1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
PubChem CID 6735
ChEBI CHEBI:17404
IUPAC Name 2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=CC=CC=C21
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10735571
View Documents Promotion Details
Thermo Scientific Acros
122110100
10 g
£27.20
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10294920
View Documents Promotion Details
Thermo Scientific Acros
122110250
25 g
£47.70
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10001220
View Documents Promotion Details
Thermo Scientific Acros
122111000
100 g
£146.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11924851
View Documents Promotion Details
Thermo Scientific Acros
122115000
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

83-33-0
132.16
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
CHEBI:17404
C1CC(=O)C2=CC=CC=C21
C9H8O
QNXSIUBBGPHDDE-UHFFFAOYSA-N
6735
2,3-dihydroinden-1-one

Specifications

83-33-0
Brown to Yellow
243.0°C to 245.0°C
Authentic
Glass bottle
07,36
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
QNXSIUBBGPHDDE-UHFFFAOYSA-N
2,3-dihydroinden-1-one
6735
132.16
Crystalline Mass
38.0°C to 42.0°C
1.1000g/mL
>110°C
99% min. (GC)
C9H8O
1.1
Solubility in water: 6.5g/L (20°C). Other solubilities: soluble in ethanol,ether and chloroform
C1CC(=O)C2=CC=CC=C21
132.16
CHEBI:17404
99+%
1-Indanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 201-470-1

Product Identifier
  • 1-Indanone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only