missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2,5-dimethoxy-4-methylbenzene, 97%, Thermo Scientific™

Product Code. 10633803
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10633803

Brand: Thermo Scientific Maybridge RH01788DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 75056-76-7
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H11IO2
MDL Number MFCD00092757
Quantity 1 g
Synonym 2,5-dimethoxy-4-iodotoluene, benzene,1-iodo-2,5-dimethoxy-4-methyl, maybridge1_006616, 1,4-dimethoxy-2-methyl-5-iodobenzene, 1-iodo-2,5-dimethoxy-4-methyl-benzene, 1-iodanyl-2,5-dimethoxy-4-methyl-benzene, 1-iodo-2,5-dimethoxy-4-methylbenzene
InChI Key BNVXMDDALQKRFP-UHFFFAOYSA-N
SMILES COC1=CC(I)=C(OC)C=C1C
IUPAC Name 1-iodo-2,5-dimethoxy-4-methylbenzene
Molecular Weight (g/mol) 278.09
PubChem CID 2728243
Formula Weight 278.08
Percent Purity 97%
Chemical Name or Material 1-Iodo-2, 5-dimethoxy-4-methylbenzene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.