missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodo-2-(trifluoromethoxy)benzene, 97%, Thermo Scientific™
Click to view available options
Quantity:
50 g
100 g
Specifications
Specifications
CAS Min % | 97.0 |
Density | 1.506g/mL |
Assay Percent Range | 97% |
Linear Formula | IC6H4OCF3 |
Quantity | 100 g |
Formula Weight | 288 |
Percent Purity | 97% |
Chemical Name or Material | 1-Iodo-2-(trifluoromethoxy)benzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'