missing translation for 'onlineSavingsMsg'
Learn More
1-Iodohexadecane, 98%, stab. with copper, Thermo Scientific Chemicals Product Code.: 15423067

1-Iodohexadecane, 98%, stab. with copper, Thermo Scientific Chemicals

Product Code. 15423067
25 g
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy

Product Code. 15423067

Brand: Thermo Scientific Alfa Aesar A12977.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from strong oxidizing agents, strong bases. Store it at a temperature of 15 - 30°C. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-77-4
Molecular Formula C16H33I
Molecular Weight (g/mol) 352.344
MDL Number MFCD00001089
InChI Key KMWHQYDMBYABKL-UHFFFAOYSA-N
Synonym cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide
PubChem CID 11007
IUPAC Name 1-iodohexadecane
SMILES CCCCCCCCCCCCCCCCI

Specifications

Melting Point 21°C to 23°C
Density 1.121
Boiling Point 206°C to 207°C (10 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)15I
Refractive Index 1.482
Quantity 25 g
Beilstein 1748533
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 352.34
Percent Purity 98%
Chemical Name or Material 1-Iodohexadecane, stabilized with copper
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244682 Certificate TypeCertificate of Analysis Date21/11/2022 Catalog NumberA12977.22,A12977.30,A12977.14
Lot NumberJ2447A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12977.22,A12977.30,A12977.14
Lot NumberJ9901A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12977.22,A12977.30,A12977.14
Lot Number10218841 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12977.22,A12977.30,A12977.14
Lot Number10188996 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12977.22,A12977.30,A12977.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Iodohexadecane, 98%, stab. with copper, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.