missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexane, 98%, stab. with copper, Thermo Scientific Chemicals

Product Code. 15453077
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-45-9
Molecular Formula C6H13I
Molecular Weight (g/mol) 212.074
MDL Number MFCD00001102
InChI Key ANOOTOPTCJRUPK-UHFFFAOYSA-N
Synonym hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane
PubChem CID 12527
IUPAC Name 1-iodohexane
SMILES CCCCCCI

Specifications

Melting Point -75°C
Density 1.437
Boiling Point 179°C to 181°C
Flash Point 61°C (141°F)
Refractive Index 1.492
Quantity 25 g
Beilstein 505971
Sensitivity Light sensitive
Formula Weight 212.08
Percent Purity 98%
Chemical Name or Material 1-Iodohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'