missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodonaphthalene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 4°C |
Density | 1.738 |
Boiling Point | 161°C to 162°C (15 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.701 |
Quantity | 10 g |
Beilstein | 1906415 |
Sensitivity | Light Sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 254.07 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'