Learn More
Hamilton™ 1 μL 7000 Series Microliter Syringe
The 7000 series syringes are modified Microliter syringes with a plunger-in-needle design.
Brand:  Hamilton™ CAL80176
 View more versions of this product
Product Code. 17804212
  • £309.00 / Each
Explore more special offers
Description
Description
  • The only zero-dead volume syringe available.
  • The fine gauge tungsten plunger wire extends to the tip of the needle, ensuring that all solvent is expelled, dispensing volumes up to 1 μL.
Specifications
Specifications
Autoclavable
23s G
0.0057 in.
0.020 in.
0.145 mm
0.515 mm
Microliter Syringe
115°C
7001 KH
1
43.4 mm
Cemented
Tungsten (Barrel)
0.0057 in.
0.145 mm
0.308 in.
7.82 mm
1.71 in.
239°F
50°F
10°C
1 μL
Cone