Learn More
Hamilton™ 1 μL 7000 Series Microliter Syringe
The 7000 series syringes are modified Microliter syringes with a plunger-in-needle design.
Brand:  Hamilton™ CAL86204
 View more versions of this product
Product Code. 17804342
  • £366.00 / Each
Explore more special offers
Description
Description
  • The only zero-dead volume syringe available.
  • The fine gauge tungsten plunger wire extends to the tip of the needle, ensuring that all solvent is expelled, dispensing volumes up to 1 μL.
Specifications
Specifications
Not Autoclavable
22 G
0.005 in.
0.028 in.
0.124 mm
0.711 mm
Microliter Syringe
115°C
7101 KHCH
1
69.9 mm
Knurled Hub, Removable
Stainless Steel
0.00490 in.
0.13 mm
0.308 in.
7.82 mm
2.75 in.
239°F
50°F
10°C
1 μL
Blunt