Learn More
Hamilton™ 1 μL 7000 Series Microliter Syringe
The 7000 series syringes are modified Microliter syringes with a plunger-in-needle design.
£266.00 - £366.00
Product Code | Brand | Needle Gauge | Needle Point Style | Connection Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Needle Point Style | Connection Type | Price | Quantity & Availability | |||
17844222
|
Hamilton™
CAL86211 |
22 G | Sharp, Bevel, Curved, Non-coring | Knurled Hub |
£266.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17804222
|
Hamilton™
CAL88400 |
22 G | Sharp, Bevel, Curved, Non-coring | Knurled Hub, Removable |
£294.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17834322
|
Hamilton™
CAL86200 |
22 G | Blunt | Knurled Hub |
£266.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17804342
|
Hamilton™
CAL86204 |
22 G | Blunt | Knurled Hub, Removable |
£366.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17804212
|
Hamilton™
CAL80176 |
23s G | Cone | Cemented |
£309.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17894202
|
Hamilton™
CAL80175 |
26s G | Cone | Cemented |
£309.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17874202
|
Hamilton™
CAL80135 |
25 G | Sharp, Bevel, Curved, Non-coring | Knurled Hub |
£307.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17884202
|
Hamilton™
CAL80100 |
25 G | Blunt | Knurled Hub |
£266.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- The only zero-dead volume syringe available.
- The fine gauge tungsten plunger wire extends to the tip of the needle, ensuring that all solvent is expelled, dispensing volumes up to 1 μL.
Specifications
115°C | |
1 | |
1 μL |