missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-butanol, Thermo Scientific Chemicals

Product Code. 16617072 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 16617072

Brand: Thermo Scientific Acros 466370050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53778-73-7
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
InChI Key CSZZMFWKAQEMPB-UHFFFAOYNA-N
IUPAC Name 1-methoxybutan-2-ol
SMILES CCC(O)COC

Specifications

Alpha Vector METHOXYBUTANOL 010
Color Colorless
Density 0.91
Boiling Point 135°C
Flash Point 39°C
Infrared Spectrum Conforms
Packaging glass bottle
Refractive Index 1.4090 to 1.4130 (20°C, 589 nm)
Quantity 5 mL
UN Number 1987
Formula Weight 104.15
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-butanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.