missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-1,2,4-triazole, 98%, Thermo Scientific Chemicals Product Code.: 11364955

1-Methyl-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Product Code. 11364955
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11364955

Brand: Thermo Scientific Alfa Aesar L18337.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6086-21-1
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.09
MDL Number MFCD01076192
InChI Key MWZDIEIXRBWPLG-UHFFFAOYSA-N
Synonym 1-methyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 1-methyl, 1-methyl,-1,2,4-triazole, 1-methyl-1h-1,2,4 triazole, pubchem22534, acmc-1avow, ksc495k1jShow MoreShow Less
PubChem CID 22459
SMILES CN1C=NC=N1

Specifications

Melting Point 16°C
Density 1.1
Boiling Point 177°C to 178°C
Flash Point 81°C (177°F)
Refractive Index 1.4705
Quantity 1 g
Beilstein 106245
IUPAC Name 1-methyl-1,2,4-triazole
Formula Weight 83.1
Percent Purity 99%
Chemical Name or Material 1-Methyl-1,2,4-triazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10147202 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL18337.06,L18337.03
Lot Number10223815 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL18337.06,L18337.03
Lot Number10108049 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL18337.06,L18337.03
Lot Number10234810 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL18337.06,L18337.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-1,2,4-triazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.