missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-4-carbaldehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 1-methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazole-4-carboxaldehyde, 1-methyl-1h-imidazole-4-carboxaldehyde, 1h-imidazole-4-carboxaldehyde, 1-methyl, methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazol-4-carboxaldehyde, 4-formyl-1-methyl-1h-imidazole, 1-methyl-4-formyl-imidazole, 1-methyl-1h-imidazole-4-carbaldehyde #, 1h-imidazole-4-carboxaldehyde,1-methyl |
Formula Weight | 110.12 |
Percent Purity | 95% |
Chemical Name or Material | 1-Methyl-1H-imidazole-4-carbaldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction