missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-3-yl)methanol, 95%, Thermo Scientific™

Catalog Number 10651704
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
84547-62-6
C5H8N2O
112.132
MFCD08690274
WCKJRVKJTUIAFW-UHFFFAOYSA-N
12845046
(1-methylpyrazol-3-yl)methanol
CN1C=CC(=N1)CO
This item is not returnable. View return policy
84547-62-6
C5H8N2O
112.132
MFCD08690274
WCKJRVKJTUIAFW-UHFFFAOYSA-N
12845046
(1-methylpyrazol-3-yl)methanol
CN1C=CC(=N1)CO
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-pyrazol-3-yl methanol, 1h-pyrazole-3-methanol, 1-methyl, 1-methylpyrazol-3-yl methanol, 1h-pyrazole-3-methanol,1-methyl, 3-hydroxymethyl-1-methyl-1h-pyrazole, 1-methylpyrazol-3-yl methan-1-ol, akos pao-1076, pubchem23752, 1-methyl-3-pyrazolyl methanol, hydroxymethyl-1-methylpyrazole
Formula Weight 112.13
Percent Purity 95%
Chemical Name or Material (1-Methyl-1H-pyrazol-3-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'