missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrrol-2-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10649505
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 69807-81-4
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD02677708
InChI Key GGCBARJYVAPZJQ-UHFFFAOYSA-N
PubChem CID 2776207
IUPAC Name (1-methylpyrrol-2-yl)methanamine
SMILES CN1C=CC=C1CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym 1-methyl-1h-pyrrol-2-yl methanamine, 1-methyl-1h-pyrrol-2-yl methylamine, 1-methylpyrrol-2-yl methanamine, 1h-pyrrole-2-methanamine, 1-methyl, 1-methyl-1h-pyrrole-2-methanamine, 1-1-methyl-1h-pyrrol-2-yl methanamine, 1-1-methylpyrrol-2-yl methanamine, 1h-pyrrole-2-methanamine,1-methyl, 1-methyl-1h-pyrrol-2-yl rnethylarnine, c-1-methyl-1h-pyrrol-2-yl methylamine
Formula Weight 110.16
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-pyrrol-2-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'